CO1H324A6CAF CombiPhos Catalysts Inc

6-Ethyl-5-bromopyridine-3-boronic acid pinacol ester

  • Cas Number : 2223036-02-8
  • Molecular Weight : 312.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart