CO1H324A6CBF CombiPhos Catalysts Inc

5-Amino-6-bromopyridine-3-boronic acid pinacol ester

  • Cas Number : 2223028-67-7
  • Molecular Weight : 298.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart