CO1H324A6CC3 CombiPhos Catalysts Inc

2-IODOPYRIDINE-4-BORONIC ACID PINACOL ESTER

  • Cas Number : 2223046-03-3
  • Molecular Weight : 330.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart