CO1H324A6CC5 CombiPhos Catalysts Inc

2-cyclobutyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)pyridine

  • Cas Number : 2223039-43-6
  • Molecular Weight : 327.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart