CO1H324A6CC8 CombiPhos Catalysts Inc

2-methyl-4-(6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)thiazole

  • Cas Number : 1402173-93-6
  • Molecular Weight : 302.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart