CO1H324A6CDA CombiPhos Catalysts Inc

3-(4,4,5,5-TETRAMETHYL-1,3,2-DIOXABOROLAN-2-YL)-5-(PIPERIDIN-4-YL)PYRIDINE

  • Cas Number : 1356066-23-3
  • Molecular Weight : 288.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart