CO1H324A6CDB CombiPhos Catalysts Inc

2-(ETHOXYCARBONYL)PYRIDINE-4-BORONIC ACID PINACOL ESTER

  • Cas Number : 741709-56-8
  • Molecular Weight : 277.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart