CO1H324A6CDD CombiPhos Catalysts Inc

2-AMINO-6-(ETHOXYCARBONYL)PYRIDINE-4-BORONIC ACID PINACOL ESTER

  • Cas Number : 2223028-97-3
  • Molecular Weight : 292.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart