CO1H324A6CDF CombiPhos Catalysts Inc

2-AMINO-6-(METHOXYCARBONYL)PYRIDINE-4-BORONIC ACID PINACOL ESTER

  • Cas Number : 2223044-78-6
  • Molecular Weight : 278.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart