CO1H324A6CE8 CombiPhos Catalysts Inc

3-(6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)benzonitrile

  • Cas Number : 2223029-71-6
  • Molecular Weight : 306.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart