CO1H324A6CEB CombiPhos Catalysts Inc

2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(m-tolyl)pyridine

  • Cas Number : 1402172-10-4
  • Molecular Weight : 295.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart