CO1H324A6CF7 CombiPhos Catalysts Inc

2-(N-Boc-Piperidin-4-yl)thiazole-5-boronic acid pinacol ester

  • Cas Number : 1884388-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart