CO1H324A6D03 CombiPhos Catalysts Inc

2-(N-Boc-piperidin-3-yl)pyrimidine-5-boronic acid pinacol ester

  • Cas Number : 1350543-60-0
  • Molecular Weight : 389.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart