CO1H324A6D0D CombiPhos Catalysts Inc

6-Bromo-5-(n-propyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223054-26-8
  • Molecular Weight : 326.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart