CO1H324A6D11 CombiPhos Catalysts Inc

6-Bromo-5-(cyclopentyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223038-91-1
  • Molecular Weight : 352.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart