CO1H324A6D16 CombiPhos Catalysts Inc

6-Chloro-5-bromopyridine-3-boronic acid pinacol ester

  • Cas Number : 2223049-72-5
  • Molecular Weight : 318.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart