CO1H324A6D1F CombiPhos Catalysts Inc

6-Chloro-5-(cyclohexyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223039-65-2
  • Molecular Weight : 321.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart