CO1H324A6D2A CombiPhos Catalysts Inc

6-Chloro-5-(4-methoxyphenyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223029-14-7
  • Molecular Weight : 345.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart