CO1H324A6D34 CombiPhos Catalysts Inc

6-Chloro-5-(4-chlorophenyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223040-12-6
  • Molecular Weight : 350.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart