CO1H324A6D46 CombiPhos Catalysts Inc

6-Chloro-5-(2-furyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223046-51-1
  • Molecular Weight : 305.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart