CO1H324A6D4D CombiPhos Catalysts Inc

6-Chloro-5-(piperidin-1-yl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223029-04-5
  • Molecular Weight : 322.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart