CO1H324A6D53 CombiPhos Catalysts Inc

6-Chloro-5-(3-methyl-1H-pyrazol-1-yl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223046-61-3
  • Molecular Weight : 319.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart