CO1H324A6D66 CombiPhos Catalysts Inc

5-Bromo-6-(cyclobutoxy)pyridine-3-boronic acid pinacol ester

  • Cas Number : 1352574-15-2
  • Molecular Weight : 354.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart