CO1H324A6D6C CombiPhos Catalysts Inc

5-Bromo-6-diethylaminopyridine-3-boronic acid pinacol ester

  • Cas Number : 1352756-86-5
  • Molecular Weight : 355.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart