CO1H324A6D6F CombiPhos Catalysts Inc

5-Bromo-6-(morpholino)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223054-48-4
  • Molecular Weight : 369.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart