CO1H324A6D70 CombiPhos Catalysts Inc

2-(1-methyl-1H-pyrazol-4-yl)-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • Cas Number : 2223029-10-3
  • Molecular Weight : 285.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart