CO1H324A6D76 CombiPhos Catalysts Inc

5-Bromo-6-(4-methyl-1H-pyrazol-1-yl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 1353877-36-7
  • Molecular Weight : 364.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart