CO1H324A6D79 CombiPhos Catalysts Inc

5-Bromo-6-(4-methylimidazol-1-yl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 1351231-47-4
  • Molecular Weight : 364.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart