CO1H324A6D81 CombiPhos Catalysts Inc

5-Bromo-6-(2-methoxyphenyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223036-44-8
  • Molecular Weight : 390.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart