CO1H324A6D89 CombiPhos Catalysts Inc

5-Bromo-6-(3-cyanophenyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223039-19-6
  • Molecular Weight : 385.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart