CO1H324A6D8D CombiPhos Catalysts Inc

5-Bromo-6-(4-trifluoromethylphenyl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223052-35-3
  • Molecular Weight : 428.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart