CO1H324A6D97 CombiPhos Catalysts Inc

5-Bromo-6-(pyridin-3-yl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223039-29-8
  • Molecular Weight : 361.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart