CO1H324A6D98 CombiPhos Catalysts Inc

5-Bromo-6-(pyridin-4-yl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223054-56-4
  • Molecular Weight : 361.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart