CO1H324A6DA7 CombiPhos Catalysts Inc

2-cyclopropyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

  • Cas Number : 2223030-90-6
  • Molecular Weight : 259.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart