CO1H324A6DAE CombiPhos Catalysts Inc

2-(4-cyclopropyl-2-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • Cas Number : 1402226-99-6
  • Molecular Weight : 312.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart