CO1H324A6DB3 CombiPhos Catalysts Inc

2-(4-chloro-3-cyclopropylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • Cas Number : 2020084-22-2
  • Molecular Weight : 278.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart