CO1H324A6DB4 CombiPhos Catalysts Inc

2-(4-bromo-3-cyclopropylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • Cas Number : 1402227-99-9
  • Molecular Weight : 323.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart