CO1H324A6DBD CombiPhos Catalysts Inc

2-(3-cyclopropyl-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • Cas Number : 627526-62-9
  • Molecular Weight : 262.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart