CO1H324A6DBE CombiPhos Catalysts Inc

2-(3-cyclopropyl-5-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • Cas Number : 1202644-19-6
  • Molecular Weight : 312.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart