CO1H324A6DC8 CombiPhos Catalysts Inc

4-cyclopropyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

  • Cas Number : 2223029-61-4
  • Molecular Weight : 269.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart