CO1H324A6DE5 CombiPhos Catalysts Inc

3-(Trifluoromethyl)-4-(4-tetrahydropyranyl)phenylboronic acid pinacol ester

  • Cas Number : 2223052-48-8
  • Molecular Weight : 356.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart