CO1H324A6DE7 CombiPhos Catalysts Inc

4,4,5,5-tetramethyl-2-(2-(tetrahydro-2H-pyran-4-yl)-4-(trifluoromethyl)phenyl)-1,3,2-dioxaborolane

  • Cas Number : 2223031-15-8
  • Molecular Weight : 356.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart