CO1H324A6E06 CombiPhos Catalysts Inc

2-(3-chloro-2-cyclopropylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • Cas Number : 2223047-25-2
  • Molecular Weight : 278.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart