CO1H324A6E0D CombiPhos Catalysts Inc

2-Cyano-6-cyclopropylphenylboronic acid pinacol ester

  • Cas Number : 2223053-37-8
  • Molecular Weight : 269.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart