CO1H324A6E10 CombiPhos Catalysts Inc

2-(2-bromo-6-cyclopropylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • Cas Number : 2223040-55-7
  • Molecular Weight : 323.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart