CO1H324A6E20 CombiPhos Catalysts Inc

1-(3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-2-methylpiperidine

  • Cas Number : 2222997-33-1
  • Molecular Weight : 319.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart