CO1H324A6E30 CombiPhos Catalysts Inc

2-(tetrahydro-2H-pyran-4-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

  • Cas Number : 2223030-23-5
  • Molecular Weight : 303.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart