CO1H324A6E3B CombiPhos Catalysts Inc

4,4,5,5-tetramethyl-2-(4-methyl-2-(tetrahydro-2H-pyran-4-yl)phenyl)-1,3,2-dioxaborolane

  • Cas Number : 2223032-06-0
  • Molecular Weight : 302.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart