CO1H324A6E58 CombiPhos Catalysts Inc

methyl 2-(tetrahydro-2H-pyran-3-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

  • Cas Number : 2222996-14-5
  • Molecular Weight : 346.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart