CO1H324A6E80 CombiPhos Catalysts Inc

2-(2-methoxy-5-(tetrahydro-2H-pyran-3-yl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • Cas Number : 2223004-40-6
  • Molecular Weight : 318.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart